Orders LinkContactUsLinkComponent
Guard columns
Guard columns
03, UNIVERSAL RP guard column for main columns with 3, 4 and 4,6 mm Ø, Guard column
目录编号 MANA728777.30
供应商:   MACHEREY-NAGEL
Guard columns
目录编号 MANA728777.30
供应商:  MACHEREY-NAGEL
Guard columns
Guard columns
03, UNIVERSAL RP guard column for main columns with 3, 4 and 4,6 mm Ø, Guard column
Jump to:
Price
询价
Log in 查看库存
1 * 3 件
Product Details & Documents
UNIVERSAL RP guard columns - to connect with the Column Protection System - are suitable for all HPLC columns under RP conditions.
  • Economical protection for analytical main column
  • Significant increase in column lifetime
规格
  • 列格式:
    Guard column
  • 说明:
    UNIVERSAL RP guard column for main columns with 3, 4 and 4,6 mm Ø
  • 顺序:
    03