Ion selective electrode, Ammonia, solid-state, IntelliCAL™
02, IntelliCAL™ ammonia ISE electrode ISENH4181, solid-state, Ag/AgCl reference system, 0,018 - 9000 mg/l NH4+-N, 0...50 °C, IntelliCAL™ ISENH4181 (ammonia), solid-state, 环氧树脂, Non refillable driTEK gel, Porous PTFE annular ring, 1 m cable, 12×220 mm
目录编号 HACHISENH4181
供应商: HACH
Product Details & Documents
IntelliCAL™ ISENH4181 is a digital combination ammonium ion selective electrode (ISE) with a non refillable double junction reference and built-in temperature sensor. The solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
- Compatible with digital Hach meters only (e.g. HQd meters)
- Provides fast, stable, and accurate response
- Ultimate traceability in measurement history
- User alerted when re-calibration is needed
- Can be moved between meters without the need to re-calibrate or re-enter measurement settings
Ideal for measuring ammonium concentrations in wastewater, drinking water and general water quality applications.
Delivery information: The electrode is supplied with calibration certificate and Basic User Manual.
The analysis package contains the IntelliCAL ISENH418101 electrode, ammonium ionic strength adjustor powder pillows (100), nitrogen-ammonia 10 mg/L standard solution (500 ml), and nitrogen-ammonia 100 mg/L standard solution (500 ml).
规格
- 浓度:0,018 - 9000 mg/l NH4+-N
- 连接:1 m cable
- 直径×长度:12×220 mm
- Type:IntelliCAL™ ISENH4181 (ammonia), solid-state
- 说明:IntelliCAL™ ammonia ISE electrode ISENH4181, solid-state, Ag/AgCl reference system
- 材质:环氧树脂
- 顺序:02
- 隔膜:Porous PTFE annular ring
- 电解质:Non refillable driTEK gel
- 温度:0...50 °C