ContactUsLinkComponent
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Catalog # MERC1.14763.0001
Supplier:  MERCK MILLIPORE
CAS Number:  
undefined
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Catalog # MERC1.14763.0001
Supplier:  MERCK MILLIPORE
CAS Number:  
Specifications
  • 化学方法:
    Koroleff digestion, dimethylphenol
  • 包装规格:
    25 Tests
  • 说明:
    Nitrogen (total) cell test
  • 顺序:
    03
  • M-Clarity quality level:
    MQ100
  • 用于单位:
    Prove, Nova 30/60, Picco
  • 范围:
    10 - 150 mg/L N
Specifications
About this item
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
ISO 23697-1: Water Quality — Determination of Total Bound Nitrogen (STTNb) in Water Using Small-Scale Sealed Tubes — Part 1
• Method: This standard describes the total nitrogen determination using the Dimethyl phenol reaction following Koroleff digestion.
• Spectroquant® Cell Tests 1.00613 and 1.14763 correspond to this norm.