



n-Heptane ≥99% (通过GC,校正水含量), UltimAR® for liquid chromatography, for organic residue analysis, 用于紫外分光光度法, Macron Fine Chemicals™
MACRV554-10
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- 包装类型:Glass bottle, amber, narrow mouth
- 包装规格:4 L
- 顺序:5162
- For Laboratory,Research,or Manufacturing Use:—
- ACS - Assay (CH₃(CH₂)₅CH₃) (by GC, corrected for water):≥99.0 %
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤1 ppm
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Water (H₂O)(by KF, coulometric):≤0.01 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 197 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:≤0.40
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:≤0.10
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 - 400 nm:≤0.01
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L):≤10 ppb
- FID Sensitive Impurities (as 2-Octano l) Single Peak (µg/L):≤5 ppb
- ACS - Neat Solvent Front Characterization:Passes Test
- Titrable Acid (µeq/g):≤0.8
:
:100.20 g/mol
:13,04679
:98,4 °C (1013 hPa)
:0,6838 g/cm³ (20 °C)
:–4 °C
:Ambient
:205-563-8
:1206
:3,II
:GHS02
:GHS08
:GHS07
:GHS09
:142-82-5
:H₃C(CH₂)₅CH₃
:MFCD00009544