Adapters, reduction, SP Wilmad-LabGlass
Catalog # LABGLG-1190-182
Supplier: LAB GLASS WILMAD
Specifications
- 圆锥体大小:34/45 NS
- 插座尺寸:24/40 NS
- 包装规格:1
Specifications
About this item
Borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Reducing adapter is used to allow connection of dissimilar NS joints. Outer joint at the top, inner joint at the bottom.