ContactUsLinkComponent
Erlenmeyer flask, with standard ground joint and stopper, SP Wilmad-LabGlass
Erlenmeyer flask, with standard ground joint and stopper, SP Wilmad-LabGlass
目录编号 LABGLG-7790-106
供应商:  LAB GLASS WILMAD
Erlenmeyer flask, with standard ground joint and stopper, SP Wilmad-LabGlass
目录编号 LABGLG-7790-106
供应商:  LAB GLASS WILMAD
规格
  • 插座尺寸:
    22 NS
  • 容量:
    125 ml
  • 包装规格:
    1
规格
关于此项目
PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Erlenmeyer flask complete with standard ground joint stopper.