Calibration weights, class 1
目录编号 TROE8144W
供应商: TROEMNER INSTRUMENTS
规格
- 重量:100 g
- 包装规格:1
- 类型:Individual weight, Class 1
规格
关于此项目
Precision-crafted of stainless steel with a highly polished finish
- Weights are straight cylinders
- Can be used as a reference standard in calibrating other weights
- Appropriate for calibrating high-precision analytical balances with a readability as low as 0,1 mg to 0,01 mg
Delivery: Supplied with a NVLAP weight calibration certificate.