VWR®, Foam Insert
Foam insert, 15 - 18 mm
目录编号 VWRU97043-590
Jump to:
Product Details & Documents
Foam insert ideal for 15 ml centrifuge tubes. Requires universal holder.
规格
- 用于管径:15 - 18 mm
- 说明:Foam insert