PYREX® Continuous Liquid/Liquid Extractor Body Assembly, Heavier Than Water, Modular
CORN3840-MCO
- 说明:Allihn condenser, medium
- 顺序:01
- 包装规格:1
Removable sidearm and crossover tube reduce chance of contamination by allowing easy, thorough cleaning
Parts are interchangeable with One-Step™ extractor/concentrators. Slightly taller form allows for longer solvent residence time and more efficient extractions. Capacity: 1000mL (34oz.).
: Sold as components only. Complete unit requires extractor body, crossover tube, sidearm, PYREX® Allihn condenser, a round-bottom flask, two [ST] 18/19 and two [ST] 35/25 ball joint clamps.