Orders LinkContactUsLinkComponent
Septa
Septa
02, Septa, 16 mm, 2 mm
目录编号 METR6.1448.020
供应商:   METROHM
Septa
目录编号 METR6.1448.020
供应商:  METROHM
Septa
Septa
02, Septa, 16 mm, 2 mm
Jump to:
Price
询价
Log in 查看库存
1 * 5 件
Product Details & Documents
These septa are made from silicone rubber.
规格
  • 直径:
    16 mm
  • 说明:
    Septa
  • 顺序:
    02
  • 厚度:
    2 mm