跳转到 页眉
跳转到 主要内容
跳转到 页脚
高级搜索
高级搜索
SDS
证书
制造商数据
0
¥0.00
Order Entry
登录
/
注册
Products
Services
Digital Solutions
Applications
Support
About
订单输入
订单状态
您的账户
购物车
China
订单输入
订单状态
您的账户
购物车
ContactUsLinkComponent
主页
色谱系统
气相色谱仪
Accessories for GC instruments
Accessories for GC instruments
REST20116
:
RESTEK
Accessories for GC instruments
REST20116
:
RESTEK
说明:
Scoring wafers, ceramic, with four straight scoring edges for cutting fused silica tubing and four serrated edges for cutting MXT® metal capillary columns, with sure grip handle
包装规格:
1