跳转到 页眉
跳转到 主要内容
跳转到 页脚
高级搜索
SDS
Certificates
制造商数据
0
¥0.00
订单输入
登录
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
您的账户
Cart
China
Order Entry
Order Status
您的账户
Cart
Orders Link
ContactUsLinkComponent
Home
缓冲(液)
酸碱缓冲液
VWR® Buffer Standard Solutions
VWR® Buffer Standard Solutions
Print
VWR® Buffer Standard Solutions
Certificates
供应商:
VWR Chemicals
VWR® Buffer Standard Solutions
供应商:
VWR Chemicals
Certificates
展开图片
Return to Top
VWR® Buffer Standard Solutions
Product Family Options
Add to cart
Jump to:
Product Family Options
关于此项目
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Documents
Certificates
按批号搜索
Product Family Options
产品信息
顺序
pH 值
pH精度
参考温度
包装类型
包装规格
可用库存
Price
VWRCE452-500ML
Certificates
01
4,00
±0,03
25 °C
Plastic bottle
500 mL
Log in
查看库存
Log in
查看库存
1 * 500 毫升
-
+
询价
1 * 500 毫升
-
+
询价
VWRCE459-500ML
Certificates
02
7,00
±0,03
25 °C
Plastic bottle
500 mL
Log in
查看库存
Log in
查看库存
1 * 500 毫升
-
+
询价
1 * 500 毫升
-
+
询价
VWRCE464-500ML
Certificates
03
10,0
±0,03
25 °C
Plastic bottle
500 mL
Log in
查看库存
Log in
查看库存
1 * 500 毫升
-
+
询价
1 * 500 毫升
-
+
询价