ContactUsLinkComponent
ICP-MS tuning solutions
ICP-MS tuning solutions
Catalog # HEWL5185-5959
CAS Number:  
undefined
ICP-MS tuning solutions
Catalog # HEWL5185-5959
CAS Number:  
Specifications
  • 浓度:
    1 μg/L
  • 基质:
    2% HNO₃
  • 包装规格:
    1
  • 说明:
    Tuning solution, 6 elements in diluted nitric acid, Li, Mg, Y, Ce, Tl, Co
  • 顺序:
    02
  • Pk Info:
    2 × 500 ml
Specifications
About this item
PA Tuning solution set includes Tuning 1 and Tuning 2 solutions.
Tuning 1: 20 ppm each of Zn, Be, Cd, As; 10 ppm each of Ni, Pb, Mg; 5 ppm each of Tl, Na, Al, U, Cu, Th, Ba, Co, Sr, V, Cr, Mn, 6Li, Sc, In, Lu, Bi; 2,5 ppm each of Y, Yb; matrix 2 to 5% HNO₃
Tuning 2: 10 ppm each of Mo, Sb, Sn, Ge, Ru, Pd; 5 ppm of Ti, Ir; matrix 10% HCl and 1% HNO₃ with trace amounts of HF.