Accessories
供应商:   BRAND
Accessories
供应商:  BRAND
Certificates
Accessories
29 Options Available Below
Product Family Options
产品信息
  • 顺序
    说明
    长度
    可用库存
    Price
  • 01
    Filling valve for Dispensette S and Dispensette S Organic 1,2,5 and 10 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 02
    Filling valve for Dispensette S and Dispensette S Organic 25, 50 and 100 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 03
    Discharge tube, without recirculation valve, shape: fine tip, 105 mm length, for Dispensette S 1,2,5 and 10 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 04
    Discharge tube, without recirculation valve, shape: standard tip, 105 mm length, for Dispensette S 5 and 10 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 05
    Discharge tube, without recirculation valve, shape: fine tip, 135 mm length, for Dispensette S 25.50 and 100 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 06
    Discharge tube, without recirculation valve, shape: standard tip, 135 mm length, for Dispensette S 25.50 and 100 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 07
    Discharge tube, without recirculation valve, shape: standard tip, 105 mm length, for Dispensette S Organic 5 and 10 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 08
    Discharge tube, without recirculation valve, shape: fine tip, 135 mm length, for Dispensette S Organic 25.50 and 100 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 09
    Discharge tube, without recirculation valve, shape: standard tip, 135 mm length, for Dispensette S Organic 25.50 and 100 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 10
    Discharge tube for Dispensette® S Trace Analysis, PT/IR valve spring, without recirculation valve, 105 mm length
    105 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 11
    Discharge tube for Dispensette® S Trace Analysis, Tantalum valve spring, without recirculation valve, 105 mm length
    105 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 12
    Discharge tube, with recirculation valve, shape: fine tip, 105 mm length, for Dispensette S 1,2,5 and 10 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 13
    Discharge tube, with recirculation valve, shape: standard tip, 105 mm length, for Dispensette S 5 and 10 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 14
    Discharge tube, with recirculation valve, shape: fine tip, 135 mm length, for Dispensette S 25.50 and 100 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 15
    Discharge tube, with recirculation valve, shape: standard tip, 135 mm length, for Dispensette S 25.50 and 100 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 16
    Discharge tube, with recirculation valve, shape: standard tip, 105 mm length, for Dispensette S Organic 5 and 10 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 17
    Discharge tube, with recirculation valve, shape: fine tip, 135 mm length, for Dispensette S Organic 25.50 and 100 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 18
    Discharge tube, with recirculation valve, shape: standard tip, 135 mm length, for Dispensette S Organic 25.50 and 100 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 19
    Discharge tube for Dispensette® S Trace Analysis, PT/IR valve spring, with recirculation valve, 105 mm length
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 20
    Discharge tube for Dispensette® S Trace Analysis, Tantalum valve spring, with recirculation valve, 105 mm length
    105 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 21
    Flexible discharge tube with recirculation valve, PTFE, coiled, length approx 800 mm, with safety handle, for Dispensette® S/S Organic 2,5+10 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 22
    Flexible discharge tube with recirculation valve, PTFE, coiled, length approx 800 mm, with safety handle, for Dispensette® S/S Organic 25,50+100 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 23
    Telescoping filling tube, FEP, adjusts to various bottle heights, length 70-140 mm for Dispensette® S/Dispensette® Organic 1,2,5 and 10 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 24
    Telescoping filling tube, FEP, adjusts to various bottle heights, length 125-240 mm for Dispensette® S/Dispensette® Organic 1,2,5 and 10 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 25
    Telescoping filling tube, FEP, adjusts to various bottle heights, length 195-350 mm for Dispensette® S/Dispensette® Organic 1,2,5 and 10 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 26
    Telescoping filling tube, FEP, adjusts to various bottle heights, length 250-480 mm for Dispensette® S/Dispensette® Organic 1,2,5 and 10 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 27
    Telescoping filling tube, FEP, adjusts to various bottle heights, length 170-330 mm for Dispensette® S/Dispensette® Organic 25,50 and 100 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 28
    Telescoping filling tube, FEP, adjusts to various bottle heights, length 250-480 mm for Dispensette® S/Dispensette® Organic 25,50 and 100 ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 29
    Dispensing cartridge for Dispensette S