Sample vials, with cap
06, Vial, VOA certified, with closed cap, 20,00 ml, 28×57 mm, 琥珀色, 是, 24-414, PTFE-faced silicone, bonded, 0,125"
目录编号 EPSCC346-0020
供应商: EP SCIENTIFIC
Jump to:
Product Details & Documents
Clear or amber sample vials made from borosilicate glass.
- Designed for water sample collection for volatile organic analysis (VOA)
- Optimised for use with automation
- With open-top white PP cap bonded with 3,17 mm thin PTFE-lined silicone septum
- Septum permanently bonded to the cap for secure sealing
- Vial shoulder slope optimized to prevent air bubble entrapment
Processed or certified to meet or exceed EPA Performance Based Specifications for volatile organic analysis with certificate. Processed vials are processed in the same manner as certified vials and come with all the features of certified vials, but vials are not barcoded and CofA is not included.
Documents
规格
- Cap included:是
- 盖尺寸:24-414
- 直径×高度:28×57 mm
- 隔膜:PTFE-faced silicone, bonded
- 容量:20,00 ml
- 颜色:琥珀色
- 说明:Vial, VOA certified, with closed cap
- 顺序:06
- 厚度:0,125"
- Cat. No.:EPSCC346-0020
- 环境认证:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact