Ion selective electrode, sodium, IntelliCAL™
HACHISENA38101
: HACH
- 浓度:0,02 - 23000 mg/l Na+
- 连接:1 m cable
- 直径×长度:12×220 mm
- Type:IntelliCAL™ ISENA381 (sodium)
- 说明:IntelliCAL™ sodium ISE electrode, Ag/AgCl reference system
- 材质:Zeonor™
- 顺序:01
- 隔膜:Refillable liquid
- 电解质:Ceramic pin
- 包装规格:1
- 温度:0...50 °C
IntelliCAL™ ISENA381 is a digital combination sodium ion selective electrode (ISE) with a refillable double junction reference and built-in temperature sensor. The probe is intended for laboratory use.
- Compatible with digital Hach meters only (e.g. HQd meters)
- Provides fast, stable, and accurate response
- Ultimate traceability in measurement history
- User alerted when re-calibration is needed
- Can be moved between meters without the need to re-calibrate or re-enter measurement settings
Ideal for measuring sodium concentrations in wastewater, drinking water and general water quality applications.
: Electrode is supplied with integrated temperature sensor, 0,02 M NH₄Cl electrolyte refill solution (60 ml), calibration certificate, and basic user manual.
The analysis package contains: IntelliCAL ISENA38101 electrode, sodium ionic strength adjustor powder pillows (100), sodium 100 mg/L standard solution (1 L), sodium 1000 mg/L standard solution (500 ml), electrode filling solution 0,02 M NH₄Cl, and dispensing cap kit.
The analysis package contains: IntelliCAL ISENA38101 electrode, sodium ionic strength adjustor powder pillows (100), sodium 100 mg/L standard solution (1 L), sodium 1000 mg/L standard solution (500 ml), electrode filling solution 0,02 M NH₄Cl, and dispensing cap kit.