ContactUsLinkComponent
Racks for sliding drawer freezer, Matrix, Matrix 2D and Nunc CryoBank
  HERA1950815
undefined
Racks for sliding drawer freezer, Matrix, Matrix 2D and Nunc CryoBank
  HERA1950815
  • 用于:
    EXF and HFU 200, 300, 400 and 600 series
  • 外部宽度×深度×高度:
    140×559×302 mm
  • 说明:
    Retangle shaped rack for 5,0 ml tubes (48 tube box)
  • 顺序:
    02
  • 包装规格:
    1
Maximise tube storage capacity per freezer with Thermo Scientific™ Sliding drawer racks for tubes (4 inner door freezers), a racking solution for four inner-door, ultra-low temperature freezers. Use in conjunction with Thermo Scientific™ dense storage vials.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Standard/deepwell microplate racks
  • Warranty varies by country
  • Material: Stainless steel
  • Colour: Silver
Thermo Scientific freezer racking and tubing solutions offer a convenient, streamlined process to maximise sample storage in your laboratory. Our racking solutions are designed to hold tubes ranging from 200 µl up to 5,0 ml, offering maximised sample storage capacity for both five and four inner door freezers.