Wipes for delicate tasks, KIMTECH SCIENCE* KIMWIPES*
目录编号 KIMB34705
供应商: KIMBERLY CLARK
规格
- 包装规格:15 boxes of 119 sheets
- 颜色:白色
- 说明:KIMTECH SCIENCE* delicate task wipes, 2 ply
- 长度×宽度:300×300 mm
- 顺序:01
规格
关于此项目
This 2-ply wipe offers a unique combination of low-lint tissue softness and multi-ply absorbency. Perfect for a multitude of delicate tasks.
- Multi-ply absorbency wipes up liquid and dust
- Wipes clean with extra low-lint and low extractable performance
- One-at-a-time dispensing