Test tubes, graduated, with beaded rim, KIMBLE®
Catalog # KIML46350-10
Supplier: KIMBLEKONTES
Specifications
- 外径:16 mm
- 容量:10 ml
- 长度:125 mm
- 包装规格:1
Specifications
About this item
Borosilicate glass, graduated.
- With beaded top
- Scale is permanent brown stain
Certifications: Conforms to USP Type I and ASTM E438, Type I, Class A requirements.