Digital block heater, with touch screen
BARN88870012
- 加热能力:600 W
- 块:4
- 温度范围:RT* +5...130 °C
- 重量:5,1 kg
- 顺序:03
- 宽度×深度×高度:535×200×100 mm
- 包装规格:1
- 温度稳定性:≤±1 °C
- 类型:Block heater, touch screen
Designed to provide precise temperature stability and uniformity. The units have an easy to use touch screen control which displays time and temperature and are durable with a powder-coated steel body. Ideal for sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation. Choice of models to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications. Interchangeable aluminum alloy sample blocks designed to provide versatility, easy cleaning and disinfecting.
- Built-in temperature sensing probe designed for outstanding temperature accuracy and control
- Temperature deviation adjustment
- Precise temperature control with PID circuit
- Temperature calibration enables user to offset the temperature to desired value
- Timer enables the user to accurately monitor the heating time
- 10 user setting programmes, each with up to 5 sequential steps of different temperatures and duration times
: Supplied with block extraction tool, but without blocks, these must be ordered separately.
: * RT = Ambient
Note: Temp. stability value shown above is at 37 °C.
Note: Temp. stability value shown above is at 37 °C.