Dropping bottles, glass, KIMBLE®
目录编号 KIML15040G-60
供应商: KIMBLEKONTES
规格
- 外径:39 mm
- 容量:60 ml
- 高度:118 mm
- 顺序:02
- 包装规格:12
规格
关于此项目
Practical, comfortable to handle universal glass dropping bottles manufactured from amber soda-lime glass conforming to USP Type III and ASTM E438, Type II requirements.
- Amber to protect from UV rays and light-sensitive substances
- Good chemical and physical properties
- For use in applications that do not involve high thermal stresses
Delivery: Supplied with 20-400 GPI screw cap, bulb and glass dropper attached on the bottle.