ContactUsLinkComponent
Capillary columns, HP-PLOT Al₂O₃ M
Capillary columns, HP-PLOT Al₂O₃ M
目录编号 HEWL19095P-M25PT
undefined
Capillary columns, HP-PLOT Al₂O₃ M
目录编号 HEWL19095P-M25PT
规格
  • 列格式:
    Capillary
  • 薄膜厚度:
    15,00 µm
  • 内径:
    0,53 mm
  • 说明:
    with two particle traps
  • 长度:
    50 m
  • 顺序:
    06
  • Phase:
    HP-PLOT Al₂O₃ M
  • 包装规格:
    1
规格
关于此项目
These columns are highly polar. Ideal for hydrocarbon analysis, and for resolving acetylene from butane and propylene from isobutane.
Aluminium oxide deactivated with proprietary deactivation