Orders LinkContactUsLinkComponent
Capillary columns, DB-17
13, on 7" cage, Capillary, DB-17, 0,25 µm, 0,25 mm, 30 m
  HEWL122-1732
Capillary columns, DB-17
  HEWL122-1732
Capillary columns, DB-17
Capillary columns, DB-17
13, on 7" cage, Capillary, DB-17, 0,25 µm, 0,25 mm, 30 m
1 * 1 件
These columns contain (50%-Phenyl)-methylpolysiloxane. They are excellent for confirmation analysis.
  • Solvent rinsable
  • Mid polarity
  • Equivalent to USP phase G3
  • Bonded and cross-linked
Max. temperature: 40 to 280/300 °C
  • 列格式:
    Capillary
  • 薄膜厚度:
    0,25 µm
  • 内径:
    0,25 mm
  • 说明:
    on 7" cage
  • 长度:
    30 m
  • 顺序:
    13
  • Phase:
    DB-17