Wipes for critical tasks, KIMTECH PREP* KIMTEX*
Catalog # KIMB33560
Supplier: KIMBERLY CLARK
Specifications
- 包装规格:66 sheets/pack
- 颜色:蓝色
- 说明:KIMTECH PREP* KIMTEX* wipes, quarterfold format
- 长度×宽度:305×318 mm
- 顺序:03
Specifications
About this item
These PP wipes are engineered for critical applications involving solvents, inks, and paints. Typical applications include: furniture finishing, engine assembly/repair, pre-packaging wipe down, LCD panel assembly, appliance assembly, printing, and glass or window cleaning.
- Acid, base and solvent resistant: Can be used with chemicals
- Perfect for use with lubricants, oil and grease
- Low linting
Colour: Blue
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904