Sampling spoons, double-ended, SP Bel-Art
Catalog # BELAH367110000
Supplier: Bel-Art Products, a Part of SP
Specifications
- 长度:229 mm
- 包装规格:1
- 线圈尺寸:13×25 mm
Specifications
About this item
Staninless steel, Fluo-Kem® PTFE coating. This spoon/spatula combination can be used for sampling and scraping.
- Corrosion resistant stainless steel construction is strong and durable
- Fluo-Kem® PTFE/FEP coating is FDA compliant and prevents material from sticking
- Spatula size: 8×51 mm
Ideal for use in compact workstations or with balances.