Floating racks for cyrotubes, Bel-Art ProCulture, SP Bel-Art
Catalog # BELAF188760020
Supplier: Bel-Art Products, a Part of SP
Specifications
- 说明:Cyrotube floating racks, 20 places, PP, Ø 95 mm
- 包装规格:1
Specifications
About this item
This 20 place polypropylene round floating rack fits most cryotubes and holds them tightly in a circular pattern formed by 12 outer and 8 inner openings.
- Floats in water; can also be used for quickly thawing frozen cells in a 37 ºC water bath
- Easily remove tubes by holding the center handle and pushing the rack down against the bench until the legs touch
- Dimensions: 6,4 mm (¹/₄") thick; 95 mm (3³/₄") in Ø; 19 mm (³/₄") long legs
- Autoclavable
Excellent for holding cryotubes during work or incubation.