Orders LinkContactUsLinkComponent
pH indicator strips
pH indicator strips
pH indicator strips, 0 - 14,0, 1
目录编号 HACH2601300
供应商:   HACH
pH indicator strips
目录编号 HACH2601300
供应商:  HACH
pH indicator strips
pH indicator strips
pH indicator strips, 0 - 14,0, 1
Jump to:
Price
询价
Log in 查看库存
1 * 100 件
Product Details & Documents
pH strips provides a simple, fast and economical method of pH measurement. Can obtain accurate results in weakly buffered, turbid and lightly coloured samples. It is easy to use and disposable.
规格
  • pH 等级:
    1
  • pH 范围:
    0 - 14,0
  • 说明:
    pH indicator strips