订单输入
China
Orders LinkContactUsLinkComponent
pH electrode storage and soaking solution

pH electrode storage and soaking solution

Electrode storage and soaking solution (4×100 ml), 1
目录编号 BECK566576
供应商:   Beckman Coulter
pH electrode storage and soaking solution
目录编号 BECK566576
供应商:  Beckman Coulter
CAS号:  
pH electrode storage and soaking solution
pH electrode storage and soaking solution

Electrode storage and soaking solution (4×100 ml), 1

Jump to:
CAS号:  
Price
询价
Log in 查看库存
Product Details & Documents

These specially formulated buffered chloride solution keeps electrodes ready for immediate use.

规格

规格

  • 包装规格:
    1
  • 说明:
    Electrode storage and soaking solution (4×100 ml)

规格