ContactUsLinkComponent
Combined ion selective electrodes, sensION
Catalog # HACH5192500
供应商:  HACH
undefined
Combined ion selective electrodes, sensION
Catalog # HACH5192500
供应商:  HACH
规格
  • 浓度:
    0,01 - saturation mg/l Na
  • 连接:
    BNC, 1 m cable
  • 直径×长度:
    12×240 mm
  • Type:
    Sodium electrode, Platinum series
  • 说明:
    Ion selective electrode, sodium
  • 顺序:
    01
  • 电解质:
    Gel (replaceable)
  • 包装规格:
    1
  • 温度:
    0...+50 °C
规格
关于此项目
These ion selective electrodes feature a replaceable gel reference electrolyte cartridge and a fixed cable with BNC connector. Compatible with Hach sensION, sensION+ and H-Series systems.
  • Replaceable gel electrolyte cartridge: Maximum life through reduced maintenance
  • Provide fast, stable, and accurate results
  • Easy to use
  • Analysis packages contain everything that is need for ISE measurements in one package
Nitrate analysis packages includes: sensION combination Platinum Series Nitrate ISE, 6-pack of replacement nitrate membrane modules, two replacement electrolyte cartridges, internal filling solution (50 ml), two Hach nitrate standard solutions (10 mg/L (500 ml) and 100 mg/L (500 ml)), Hach nitrate ionic strength adjustor (ISA) Powder Pillows (pack of 100), 50-ml beaker and ISE User Manual.
Delivery: Electrodes are supplied with two replacement electrolyte cartridges and user manual.