PCR superMix, SsoFast™ EvaGreen®
Catalog # BRAD172-5211
Supplier: BIO RAD
CAS Number:
Specifications
- 包装规格:5 mL
- Application:qPCR dye
- 说明:PCR supermix, SsoFast™ EvaGreen®, with low ROX, 500×20 µl reactions
- 顺序:02
Specifications
About this item
SsoFast™ EvaGreen® PCR Supermix with low ROX is a 2X concentrated, ready to use reaction master mix containing all components except primers and template for real time quantitative PCR on ROX-dependent real time PCR instruments. It contains antibody mediated Sso7d-fusion polymerase, dNTPs, MgCl₂, EvaGreen dye, enhancers, stabilisers and ROX.
- Fast qPCR results under fast cycling conditions (approximately 40 min)
- Preblended with ROX
- Minimal PCR inhibition provides better performance with fast cycling protocols
- Single copy detection of target genes