ContactUsLinkComponent
Stoppers for conical ground joints, PYREX®
Stoppers for conical ground joints, PYREX®
Catalog # CORN7570N-24
undefined
Stoppers for conical ground joints, PYREX®
Catalog # CORN7570N-24
Specifications
  • 模拟器:
    24/40
  • 说明:
    Reagent bottle stoppers with hollow glass standard taper joint
  • 顺序:
    02
  • 包装规格:
    6
Specifications
About this item
Hollow, full length stopper closed at the bottom from PYREX®, borosilicate glass.
  • Interchangeable with standard taper joints
They are lightweight, but strong enough to keep chipping and breakage to a minimum. Barrel-shaped head is easy to clean and rotate, side grooves prevent slippage, and flat top allows stopper to stand on head.