Solution bottles, graduated, PYREX®
证书
关于此项目
PYREX® borosilicate glass.
- With double scale (liters and gallons) black enamel graduations
- Graduated in 500 ml increments
- Will withstand hot air (dry) or steam (wet) sterilisation
- Necks are tooled to increase mechanical strength and for uniform stopper fit
Solution bottles are designed for storage of solutions and are ideal for media preparation.