Dropper tips, WHEATON®
71, 绿色
WHEAW242436-A
LDPE, coloured.
- Extended controlled dropper tips
- Dispense one drop at a time
- Average drop size is 44 ±6 μl
Ordering information: Tips available in different colours and packaged in case of 100 or 1000.
- 颜色:绿色
- 顺序:71