Orders LinkContactUsLinkComponent
PBS buffer
01, PBS buffer 10X, includes 0,1 M phosphate buffer with 27 mM KCl and 1,37 M NaCl, 1.000 mL
目录编号 GEHEBR-1006-72
供应商:   GE HEALTHCARE
PBS buffer
目录编号 GEHEBR-1006-72
供应商:  GE HEALTHCARE
CAS号:  
PBS buffer
PBS buffer
01, PBS buffer 10X, includes 0,1 M phosphate buffer with 27 mM KCl and 1,37 M NaCl, 1.000 mL
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 1.000 毫升
Product Details & Documents
Phosphate buffered saline solution, available with or without detergent, recommended as running buffer for small molecule analysis in Biacore systems. Concentrated stock solution, yielding pH 7,4 when diluted 10X.
  • Can be stored at room temperature, precipitation can occur at low temperature
  • Supplemented with DMSO
规格
规格
  • 包装规格:
    1.000 mL
  • 说明:
    PBS buffer 10X, includes 0,1 M phosphate buffer with 27 mM KCl and 1,37 M NaCl
  • 顺序:
    01
  • Cat. No.:
    GEHEBR-1006-72
  • 用于:
    Interaction analyses in Biacore systems except Biacore Flexchip
规格