Specifications
- 容量:6 ml
- 工作台重量:100 mg
- Phase:Hydrophobic SC-DVB
- 包装规格:1
Specifications
About this item
These columns feature BAKERBOND® DVB polymer resins, which are the product of ultra-clean polymer microparticle technology. These resin particles have a large surface area, are highly rigid, are stable over pH range 1 to 14, are water-wettable and are not impacted by sorbent drying. These columns are recommended when advanced detection methods are used.
- Improve sample preparation recovery
- Polymer based
General applications: Ionic, basic analytes.
These SPE columns can be used in mixed mode.