Specifications
- 颜色:蓝色
- 说明:MegaSorb™ mop head, 10 oz., bulk pack
- 材质:超细纤维
- 顺序:01
- Environmentally Preferable:Waste Reducing
- 包装规格:1
Specifications
About this item
A highly absorbent mop that combines the cleaning efficiency of microfibre with the ease of a looped string mop. The tufted strands are comprised of polyester and nylon filaments, designed to pick up liquids and tiny particles in controlled and critical environments.
- Highly absorbent and low-linting
- Polyester and nylon tufted microfibre tubes
- Picks up tiny particles where cotton fibres just roll over
- White microfibre option includes an additional scrubbing band
- Use with MH-1 handle or QDMH-1 MicroMop adapter with SSU series handles
- Compatible with alcohol and strong disinfectants
- Launderable for reuse
- Autoclavable
The M-Zone™ MegaSorb™ Microfibre String Mop features distinctive microfiber tubes with tufted strands made of polyester and nylon filaments, 30 times smaller than cotton fibres, that work to pick up tiny particles.