Rapid Resolution High Definition (RRHD) columns, ZORBAX®
HEWL863953-912
- 列格式:Analytical
- 内径:4,60 mm
- 颗粒大小:1,8 µm
- 孔大小:80 Å
- 长度:150 mm
- 顺序:26
- Phase:Phenyl
- 包装规格:1
These columns are ideal for fast or high-resolution separations. Greater flexibility that allows to push flow rates to the limit without compromising the efficiency or quality of the separations.
- Stability up to 1200 bar, for ultimate compatibility with UHPLC instruments
- Use ZORBAX® Eclipse Plus to achieve the exceptionally symmetrical peaks that deliver reliable results
- Use ZORBAX® SB-C18 for different selectivity and enhanced stability at pH 1 to 2
- More reliable compound identification due to high definition chromatograms and 2,1 mm ID columns that enhance MS
- Ensured scalability and method transfer