ContactUsLinkComponent
Analytical HPLC columns, XBridge™ BEH Shield RP18
Analytical HPLC columns, XBridge™ BEH Shield RP18
目录编号 WATE186003045
供应商:  WATERS
Analytical HPLC columns, XBridge™ BEH Shield RP18
目录编号 WATE186003045
供应商:  WATERS
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
规格
  • 列格式:
    Analytical
  • 内径:
    4,60 mm
  • 颗粒大小:
    3,5 µm
  • 孔大小:
    130 Å
  • 长度:
    150 mm
  • 顺序:
    04
  • Phase:
    BEH Shield RP18
  • 包装规格:
    1
规格
关于此项目
XBridge™ BEH Shield RP18 columns complement the XBridge™ BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.