Analytical HPLC columns, SymmetryShield™ RP18
目录编号 WATE186000180
供应商: WATERS
规格
- 内径:4,60 mm
- 颗粒大小:3,5 µm
- 孔大小:100 Å
- 长度:150 mm
- 顺序:03
- Phase:RP18
- 包装规格:1
规格
关于此项目
SymmetryShield™ columns feature Waters' patented embedded polar group technology that shields the silica's residual silanols from highly basic analytes at improves overall peak shape. Additionally, by placing the embedded polar group close to the silica surface, the activity of the surface silanols is further reduced. This imparts selectivity and retention that is different compared to the Symmetry® C18 ligand.