PCR Plates, 96-well, low profile, Multiplate™
目录编号 BRADMLL-9601
供应商: BIO RAD
规格
- 孔数量:96
- Profile:低
- 裙边:无裙边
- Well colour:透明
- 包装规格:25
规格
关于此项目
Multiplate™ PCR plates are compatible with many instruments and a wide variety of sealing systems including all Bio-Rad PCR Plate Seals and the PX1™ PCR Plate Sealer.
- Low overall height (15,50 mm) reduces condensation
- Compatible with optical real-time PCR detection systems
- Easily cut with scissors when less than a full plate is needed
- Free of DNase, RNase, and human DNA
These PP plates are ideal for fast PCR and low-volume reactions (5 – 125 µl; 200 µl max per well).