ContactUsLinkComponent
PCR Plates, 96-well, low profile, Multiplate™
PCR Plates, 96-well, low profile, Multiplate™
目录编号 BRADMLL-9601
供应商:  BIO RAD
PCR Plates, 96-well, low profile, Multiplate™
目录编号 BRADMLL-9601
供应商:  BIO RAD
规格
  • 孔数量:
    96
  • Profile:
  • 裙边:
    无裙边
  • Well colour:
    透明
  • 包装规格:
    25
规格
关于此项目
Multiplate™ PCR plates are compatible with many instruments and a wide variety of sealing systems including all Bio-Rad PCR Plate Seals and the PX1™ PCR Plate Sealer.
  • Low overall height (15,50 mm) reduces condensation
  • Compatible with optical real-time PCR detection systems
  • Easily cut with scissors when less than a full plate is needed
  • Free of DNase, RNase, and human DNA
These PP plates are ideal for fast PCR and low-volume reactions (5 – 125 µl; 200 µl max per well).