Injection port liners for Agilent Technologies instruments
HYPE453A1925
- 外径:6,3
- 内径:4
- 说明:Splitless liner with single taper, quartz wool packing
- 长度:78,5 mm
- 顺序:21
- 包装规格:5
Optimise the performance of Agilent™ GC systems with these specifically designed injection port liners for Agilent instruments.
- Highly deactivated
- Produced to exacting tolerances
- Highly deactivated borosilicate glass