Bench top protector sheets, KIMTECH SCIENCE*
Catalog # KIMB75450
Supplier: KIMBERLY CLARK
Specifications
- 包装规格:Roll
- 颜色:白色/绿色
- 长度×宽度:486×76 200 mm
- 顺序:02
Specifications
About this item
This advanced bench top protector made from Airlaid material features a polyethylene backing for excellent slip resistance.
- Quickly absorbs spills
- Flat, stable surface helps protect experiments and delicate glassware
- Remains flat, even after drying