Microplates, 384-well, non binding
Catalog # GREI781901_40
Supplier: GREINER BIO ONE
Specifications
- 颜色:透明
- 说明:384-well microplate, non binding, flat bottom
- 顺序:02
- Environmentally Preferable:Low Manufacturing Impact
- 环境认证:ISO 14001 (241002); ISO 50001 (241002)
- 包装规格:40
Specifications
About this item
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
All plates are free from RNase, DNase, and pyrogens, and are also free of DNA polymerase inhibitors.