ContactUsLinkComponent
VWR® Lysis Buffer, Hypotonic
VWR® Lysis Buffer, Hypotonic
目录编号 VWRCM334-100ML
供应商:  VWR Chemicals
CAS 编号:  
undefined
VWR® Lysis Buffer, Hypotonic
目录编号 VWRCM334-100ML
供应商:  VWR Chemicals
CAS 编号:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
规格
  • 包装类型:
    Plastic bottle
  • 包装规格:
    100 mL
  • 说明:
    Hyptonic lysis buffer
规格
关于此项目
Detergent-free buffer for isolation of proteins from tissue culture cells. Cell lysis and the subsequent isolation of the crude cytoplasmic fraction completed in less than 30 m.
The protocol is compatible with a variety of downstream applications including Western Blotting and ELISA procedures, as well as protein assays, which are sensitive to the presence of detergents in the lysis buffer.