Orders LinkContactUsLinkComponent
Test kits, digital titration method
供应商:   HACH
Test kits, digital titration method
供应商:  HACH
Certificates
Test kits, digital titration method
Test kits, digital titration method
22 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Product Family Options
Test kits with titrator
产品信息
  • 顺序
    说明
    范围
    包装规格
    可用库存
    Price
  • 06
    Total hardness HA-DT: EDTA method
    10 - 4000 mg/l
    1 KIT
    Log in 查看库存
    Log in 查看库存
    1 * 1 KIT
    1 * 1 KIT
Reagent sets with digital titration cartridge (without titrator)
产品信息
  • 顺序
    说明
    范围
    包装规格
    可用库存
    Price
  • 07
    Chloride reagent kit: Mercury nitrate method
    10 - 8000 mg/l
    100 Tests
    Log in 查看库存
    Log in 查看库存
    1 * 100 Tests
    1 * 100 Tests
  • 08
    Chloride reagent kit: silver nitrate method
    10 - 10000 mg/l
    1 KIT
    Log in 查看库存
    Log in 查看库存
    1 * 1 KIT
    1 * 1 KIT
  • 11
    Chloride testing: 1,5-diphenyl carbazone powder pillows by mercuric nitrate titration method
    10 - 8000  mg/l
    100
    Log in 查看库存
    Log in 查看库存
    1 * 100 件
    1 * 100 件
  • 13
    Hardness (Calcium) reagent set, high range
    10 - 4000 mg/l
    100 Tests
    Log in 查看库存
    Log in 查看库存
    1 * 100 Tests
    1 * 100 Tests
  • 14
    Hardness (Calcium) reagent set, low range
    10 - 160 mg/l
    1
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 16
    Total hardness LR: EDTA method
    10 - 160 mg/l
    100 Tests
    Log in 查看库存
    Log in 查看库存
    1 * 100 Tests
    1 * 100 Tests
  • 17
    Total hardness HR: EDTA method
    100 - 4000 mg/l
    100 Tests
    Log in 查看库存
    Log in 查看库存
    1 * 100 Tests
    1 * 100 Tests
  • 20
    Iron (Total) reagent set, high range
    100 - 1000 mg/l
    1
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 21
    Sulphite reagent kit: iodate-iodide method
    4 - 400 mg/l
    100 Tests
    Log in 查看库存
    Log in 查看库存
    1 * 100 Tests
    1 * 100 Tests
Digital titration cartridges/kits (without titrator)
产品信息
  • 顺序
    说明
    范围
    包装规格
    可用库存
    Price
  • 23
    Sulfuric acid digital titrator cartridge, 0,1600 N
    13 mL
    Log in 查看库存
    Log in 查看库存
    1 * 13 毫升
    1 * 13 毫升
  • 25
    Sulfuric acid digital titrator cartridge, 8,00 N
    1
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 26
    Mercuric nitrate* 0,2256 N cartridge
    1
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 27
    Silver nitrate digital titrator cartridge, 0,2256 N
    1
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 28
    Silver nitrate digital titrator cartridge, 1,128 N
    1
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 30
    Cyclohexanediamine tetraacetic acid digital titrator cartridge, 0,0800 M
    1
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 31
    Cyclohexanediamine tetraacetic acid digital titrator cartridge, 0,800 M
    1
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 32
    Digital titrator cartridge assembly, empty
    1
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 33
    Sulphite: iodate-iodide 0.3998 N cartridge
    1
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 39
    Ferrous ethylenediammonium sulphate digital titrator cartridge, 0,00564 N
    1
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 41
    Sodium thiosulphate (Stabilised) digital titrator cartridge, 2,26 N
    1
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 42
    Mercuric nitrate* 2,256 N
    13 mL
    Log in 查看库存
    Log in 查看库存
    1 * 13 毫升
    1 * 13 毫升