Orders LinkContactUsLinkComponent
Test kits, drop count tests
Test kits, drop count tests
供应商:   HACH
Test kits, drop count tests
供应商:  HACH
Certificates
Test kits, drop count tests
Test kits, drop count tests
9 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Product Family Options
Test kits
产品信息
  • 顺序
    说明
    范围
    每套测试
    包装规格
    可用库存
    Price
  • 04
    Carbon dioxide, model CA-23
    1,25 - 25, 2 - 40 and 5 - 100 mg/l
    1 KIT
    Log in 查看库存
    Log in 查看库存
    1 * 1 KIT
    1 * 1 KIT
  • 05
    Chloride, model 8-P, use silver nitrate titrant
    5 - 100 mg/l Cl-
    20 - 400 mg/l Cl-
    1 KIT
    Log in 查看库存
    Log in 查看库存
    1 * 1 KIT
    1 * 1 KIT
  • 13
    Hydrogen peroxide, model HYP-1
    1 - 10 mg/l
    1 KIT
    Log in 查看库存
    Log in 查看库存
    1 * 1 KIT
    1 * 1 KIT
  • 15
    Sulphiite, model SU-5
    1 KIT
    Log in 查看库存
    Log in 查看库存
    1 * 1 KIT
    1 * 1 KIT
  • 16
    Total hardness, model 5-B
    17 - 510 mg/l
    1 KIT
    Log in 查看库存
    Log in 查看库存
    1 * 1 KIT
    1 * 1 KIT
  • 17
    Total hardness, model 5-EP
    17 - 510 mg/l
    100
    1 KIT
    Log in 查看库存
    Log in 查看库存
    1 * 1 KIT
    1 * 1 KIT
  • 18
    Total hardness, model HA-71A
    1 - 20 mg/l
    1 KIT
    Log in 查看库存
    Log in 查看库存
    1 * 1 KIT
    1 * 1 KIT
Replacement reagent set
产品信息
  • 顺序
    说明
    范围
    每套测试
    包装规格
    可用库存
    Price
  • 22
    Hardness 3 solution, 100 ml MDB
    100 mL
    Log in 查看库存
    Log in 查看库存
    1 * 100 毫升
    1 * 100 毫升
  • 25
    Sulfite 3 reagent, 100 ml MDB
    100 mL
    Log in 查看库存
    Log in 查看库存
    1 * 100 毫升
    1 * 100 毫升