Centrifuge tube, round bottom, with screw cap, KIMAX®, KIMBLE®
KIML45212-50
- 盖:配有螺帽
- 直径×长度:29×123 mm
- 包装规格:12/case
- 螺纹:24-410
- 容量:50 ml
- 长度:123 mm
Tube has a heavy wall to withstand higher speeds in centrifuging.
It is especially useful in separating neutralised sediment for culturing from sputum specimens, as well as many other clinical and microbiological applications.
: Supplied with unattached autoclavable cap with cemented-in rubber liner.