Measuring (Mohr) pipettes, color-coded, tempered top and tip, colored markings, PYREX®
目录编号 CORN7065-10
规格
- 颜色代码:橙色
- 容量:10 ml
- 长度:380 mm
- 顺序:06
- 分度:0,1 ml
- 包装规格:12
- 误差:±0,06 ml
规格
关于此项目
These reusable glass pipettes are colour-coded by size for easy identification and sorting.
- Top end is constricted (Mohr type)
- Coloured graduations are enamelled onto the glass
- Calibrated 'to deliver' (TD) their total capacity without blow-out
Sharp, fine lines provide accurate reading. Large numbers are easy to read. The top end of 5 to 25 ml sizes is constricted.
Certifications: ASTM E-1293