Solution bottles, PYREX®
CORN1595-2X
- 容量:9,5 L
- 说明:Solution Bottle with Tooled Neck
- 顺序:01
- 包装规格:4
These bottles are designed for storage of solutions and are ideal for media preparation.
The 9.5 liter (21/2 gallon) and 13.25 liter (31/2 gallon) sizes are in a conventional bottle shape. The 19 liter (5 gallon) and 45.5 liter (12 gallon) sizes are similar in design to a carboy. The necks are tooled to increase mechanical strength and for uniform stopper fit.