Serum bottles, with tooled neck, PYREX®
目录编号 CORN1585-4L
规格
- 外径:165 mm
- 容量:4000 ml
- 颜色:透明
- 高度:278 mm
- 顺序:01
- 管颈外径:49 mm
- 包装规格:1
规格
关于此项目
Designed for the handling and storage of sterile culture media and sera when the stability of glass is of prime importance.
- Fits a #8 rubber stopper
- Stopper not included
- Bottle withstands repeated wet or dry autoclaving
These serum bottles are made of Pyrex® borosilicate glass.
Also suitable for storing distilled water and standard solutions. Necks are tooled for uniform rubber stopper fit and increased mechanical strength.